siRNA Details
| Gene Symbol: FAM19A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005527.2 | NP_001005527.1 | 3 | 485 | ||
| NM_182522.4 | NP_872328.1 | 3 | 567 | |||
| XM_011533371.1 | XP_011531673.1 | 3 | 567 | |||
| XM_011533372.1 | XP_011531674.1 | 3 | 443 | |||
| GenBank mRNA | AK057890.1 | - | 2 | 183 | ||
| AY325117.1 | - | 3 | 539 | |||
| BC031566.1 | - | 3 | 399 | 5195201 | ||

FAM19A4
Genomic Map