siRNA Details
Gene Symbol: XXYLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308069.1 | NP_001294998.1 | 4 | 641 | ||
NM_152531.4 | NP_689744.3 | 4 | 1104 | |||
XM_005269286.4 | XP_005269343.1 | 4 | 3489 | |||
XM_011512448.2 | XP_011510750.1 | 5 | 916 | |||
XM_017005750.1 | XP_016861239.1 | 3 | 3877 | |||
GenBank mRNA | AK057046.1 | - | 5 | 849 | ||
AK075551.1 | - | 4 | 1072 | |||
AK092474.1 | - | 2 | 381 | |||
AK301133.1 | - | 4 | 403 | |||
BC019036.2 | - | 4 | 1013 | 4591786 | ||
BC039067.1 | - | 4 | 1086 | 5185557 | ||
BG721419.1 | - | 4 | 646 |
