siRNA Details
| Gene Symbol: CMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182523.1 | NP_872329.1 | 3 | 302 | ||
| XM_011533402.2 | XP_011531704.1 | 5 | 479 | |||
| XM_017005760.1 | XP_016861249.1 | 4 | 261 | |||
| GenBank mRNA | AF086095.1 | - | 1 | 464 | ||
| BC052644.1 | - | 3 | 302 | 6163781 | ||
| BX648671.1 | - | 3 | 329 | |||
| CR749370.1 | - | 3 | 352 | |||

CMC1
Genomic Map