siRNA Details
| Gene Symbol: CMTM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178868.4 | NP_849199.2 | 2 | 516 | ||
| XM_011533416.2 | XP_011531718.1 | 3 | 316 | |||
| XM_017005779.1 | XP_016861268.1 | 2 | 836 | |||
| XM_017005780.1 | XP_016861269.1 | 2 | 190 | |||
| GenBank mRNA | AF474370.2 | - | 2 | 516 | ||

CMTM8
Genomic Map