siRNA Details
| Gene Symbol: CMTM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320308.1 | NP_001307237.1 | 2 | 481 | ||
| NM_178868.4 | NP_849199.2 | 3 | 655 | |||
| XM_011533416.2 | XP_011531718.1 | 4 | 455 | |||
| XM_017005779.1 | XP_016861268.1 | 3 | 975 | |||
| XM_017005780.1 | XP_016861269.1 | 3 | 329 | |||
| GenBank mRNA | AF474370.2 | - | 3 | 655 | ||
| AY598783.1 | - | 2 | 481 | |||
| BC041390.2 | - | 3 | 610 | 5276695 | ||
| CB070190.1 | - | 1 | 46 | |||

CMTM8
Genomic Map