siRNA Details
| Gene Symbol: C4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152543.2 | NP_689756.2 | 4 | 592 | ||
| XM_011531671.2 | XP_011529973.1 | 5 | 627 | |||
| XM_011531672.2 | XP_011529974.1 | 5 | 737 | |||
| XM_017007808.1 | XP_016863297.1 | 8 | 1493 | |||
| XM_017007809.1 | XP_016863298.1 | 8 | 1423 | |||
| XM_017007810.1 | XP_016863299.1 | 8 | 1599 | |||
| GenBank mRNA | AK058100.1 | - | 4 | 592 | ||
| AK307319.1 | - | 3 | 294 | |||
| DB037602.1 | - | 3 | 294 | |||

C4orf45
Genomic Map