siRNA Details
| Gene Symbol: TRMT44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152544.2 | NP_689757.2 | 7 | 1268 | ||
| XM_005247950.3 | XP_005248007.1 | 7 | 1265 | |||
| XM_011513406.2 | XP_011511708.1 | 7 | 1297 | |||
| XM_011513407.2 | XP_011511709.1 | 7 | 1297 | |||
| XM_011513408.1 | XP_011511710.1 | 8 | 1409 | |||
| XM_011513409.2 | XP_011511711.1 | 8 | 1380 | |||
| GenBank mRNA | AK093044.1 | - | 3 | 274 | ||
| BC035655.1 | - | 7 | 768 | 5588410 | ||

TRMT44
Genomic Map