siRNA Details
| Gene Symbol: ARSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198150.2 | NP_937793.1 | 8 | 1557 | ||
| XM_005271904.3 | XP_005271961.1 | 9 | 1621 | |||
| GenBank mRNA | AK303855.1 | - | 8 | 1609 | ||
| AL832711.1 | - | 7 | 1240 | |||
| AY358596.1 | - | 8 | 1517 | |||
| AY875939.1 | - | 8 | 1399 | |||
| BC036047.1 | - | 8 | 1419 | |||
| BC050392.1 | - | 9 | 1567 | |||
| BC130329.1 | - | 8 | 1495 | 40146360 | ||
| BC144161.1 | - | 9 | 1554 | |||

ARSK
Genomic Map