siRNA Details
| Gene Symbol: ARSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198150.2 | NP_937793.1 | 4 | 641 | ||
| XM_005271904.3 | XP_005271961.1 | 5 | 705 | |||
| GenBank mRNA | AK303855.1 | - | 4 | 693 | ||
| AK316253.1 | - | 4 | 661 | |||
| AL832711.1 | - | 3 | 324 | |||
| AY358596.1 | - | 4 | 601 | |||
| AY875939.1 | - | 4 | 483 | |||
| BC036047.1 | - | 4 | 503 | |||
| BC050392.1 | - | 5 | 651 | |||
| BC130329.1 | - | 4 | 579 | 40146360 | ||
| BC144161.1 | - | 4 | 579 | |||

ARSK
Genomic Map