siRNA Details
| Gene Symbol: FAM81B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152548.2 | NP_689761.2 | 9 | 1122 | ||
| XM_005271905.2 | XP_005271962.2 | 8 | 981 | |||
| XM_005271906.4 | XP_005271963.2 | 11 | 1286 | |||
| XM_011543209.1 | XP_011541511.1 | 7 | 803 | |||
| XM_011543210.2 | XP_011541512.1 | 6 | 836 | |||
| XM_017009118.1 | XP_016864607.1 | 10 | 1142 | |||
| GenBank mRNA | AK058062.1 | - | 9 | 1105 | ||
| AK310245.1 | - | 9 | 1065 | |||
| BC034772.1 | - | 9 | 1122 | 5169904 | ||

FAM81B
Genomic Map