siRNA Details
| Gene Symbol: FAM71B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130899.2 | NP_570969.2 | 2 | 924 | ||
| GenBank mRNA | AK057543.1 | - | 2 | 920 | ||
| BC022035.1 | - | 2 | 932 | 4826350 | ||
| BC025397.1 | - | 2 | 925 | 4827541 | ||
| BC025409.1 | - | 2 | 925 | 4825198 | ||
| BC025998.1 | - | 2 | 925 | 4836822 | ||

FAM71B
Genomic Map