siRNA Details
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144726.2 | NP_653327.1 | 1 | 43 | ||
GenBank mRNA | AK056513.1 | - | 1 | 43 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144726.2 | NP_653327.1 | 1 | 43 | ||
GenBank mRNA | AK056513.1 | - | 1 | 43 |