siRNA Details
| Gene Symbol: SNRNP48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152551.3 | NP_689764.3 | 8 | 959 | ||
| XM_011514312.2 | XP_011512614.1 | 8 | 895 | |||
| GenBank mRNA | AK027220.1 | - | 3 | 300 | ||
| AK056796.1 | - | 8 | 2598 | |||
| AK292409.1 | - | 8 | 937 | |||
| AL832462.1 | - | 8 | 960 | |||
| BC062458.1 | - | 8 | 921 | |||
| BC114630.1 | - | 8 | 927 | 40084896 | ||
| BC114631.1 | - | 8 | 928 | 40084903 | ||

SNRNP48
Genomic Map