siRNA Details
| Gene Symbol: SLC2A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207420.2 | NP_997303.2 | 12 | 1427 | ||
| GenBank mRNA | AY571960.2 | - | 12 | 1427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC2A7| Gene Symbol: SLC2A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207420.2 | NP_997303.2 | 12 | 1427 | ||
| GenBank mRNA | AY571960.2 | - | 12 | 1427 | ||