siRNA Details
Gene Symbol: SLC2A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207420.2 | NP_997303.2 | 4 | 326 | ||
XM_011540824.2 | XP_011539126.1 | 4 | 365 | |||
XM_011540825.2 | XP_011539127.1 | 4 | 365 | |||
GenBank mRNA | AY571960.2 | - | 4 | 326 |
