siRNA Details
| Gene Symbol: FER1L6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103547.1 | 2 | 507 | |||
| GenBank mRNA | AK057332.1 | - | 2 | 507 | ||
| BC101502.1 | - | 2 | 448 | |||
| BC101506.1 | - | 2 | 448 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FER1L6-AS2| Gene Symbol: FER1L6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103547.1 | 2 | 507 | |||
| GenBank mRNA | AK057332.1 | - | 2 | 507 | ||
| BC101502.1 | - | 2 | 448 | |||
| BC101506.1 | - | 2 | 448 | |||