siRNA Details
| Gene Symbol: FAM84B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174911.4 | NP_777571.1 | 2 | 1285 | ||
| XM_017013107.1 | XP_016868596.1 | 2 | 1332 | |||
| XM_017013108.1 | XP_016868597.1 | 3 | 1264 | |||
| GenBank mRNA | AJ417849.1 | - | 2 | 1212 | ||
| AK292824.1 | - | 2 | 1210 | |||
| BC052957.1 | - | 2 | 1040 | 5262890 | ||

FAM84B
Genomic Map