siRNA Details
| Gene Symbol: TDRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175075.4 | NP_778250.2 | 4 | 780 | ||
| XM_006716232.3 | XP_006716295.1 | 3 | 760 | |||
| XM_017013113.1 | XP_016868602.1 | 3 | 1094 | |||
| GenBank mRNA | AK126209.1 | - | 1 | 245 | ||
| AY194292.2 | - | 3 | 598 | |||
| BC136248.1 | - | 3 | 759 | 9020235 | ||
| BC136249.1 | - | 3 | 759 | 9020236 | ||
| BX648606.1 | - | 2 | 383 | |||
| CN430635.1 | - | 3 | 591 | |||

TDRP
Genomic Map