siRNA Details
| Gene Symbol: SBSPON | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153225.3 | NP_694957.3 | 3 | 662 | ||
| XM_017013145.1 | XP_016868634.1 | 3 | 907 | |||
| GenBank mRNA | AK292970.1 | - | 3 | 562 | ||
| AY040546.1 | - | 3 | 560 | |||
| BC042877.1 | - | 3 | 662 | 5169941 | ||

SBSPON
Genomic Map