siRNA Details
| Gene Symbol: CAMSAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015447.3 | NP_056262.3 | 11 | 2097 | ||
| XM_005263396.3 | XP_005263453.1 | 12 | 2522 | |||
| XM_005263397.1 | XP_005263454.1 | 9 | 1623 | |||
| XM_011518272.1 | XP_011516574.1 | 11 | 1791 | |||
| XM_011518273.2 | XP_011516575.1 | 10 | 1622 | |||
| XM_011518274.1 | XP_011516576.1 | 9 | 1652 | |||
| XM_017014301.1 | XP_016869790.1 | 11 | 2378 | |||
| XM_017014302.1 | XP_016869791.1 | 11 | 2302 | |||
| GenBank mRNA | AJ519841.1 | - | 9 | 1622 | ||
| AL117634.1 | - | 1 | 627 | |||
| BC012778.2 | - | 1 | 97 | |||
| BC130580.1 | - | 11 | 1756 | 40146611 | ||
| BC144082.1 | - | 11 | 1756 | 9052602 | ||

CAMSAP1
Genomic Map