siRNA Details
| Gene Symbol: C9orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152571.2 | NP_689784.1 | 1 | 1266 | ||
| GenBank mRNA | AK055336.1 | - | 1 | 1218 | ||
| AK094098.1 | - | 1 | 537 | |||
| BC036221.1 | - | 1 | 1266 | |||
| BC117152.1 | - | 1 | 370 | 40125703 | ||
| BC117154.1 | - | 1 | 370 | 40125705 | ||

C9orf163
Genomic Map