siRNA Details
| Gene Symbol: MAMDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206920.2 | NP_996803.2 | 18 | 2355 | ||
| GenBank mRNA | AL137659.1 | - | 17 | 2898 | ||
| AL834531.1 | - | 3 | 370 | |||
| AY358419.1 | - | 18 | 2355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAMDC4| Gene Symbol: MAMDC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206920.2 | NP_996803.2 | 18 | 2355 | ||
| GenBank mRNA | AL137659.1 | - | 17 | 2898 | ||
| AL834531.1 | - | 3 | 370 | |||
| AY358419.1 | - | 18 | 2355 | |||