siRNA Details
| Gene Symbol: AK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317958.1 | NP_001304887.1 | 12 | 1730 | ||
| NM_001317959.1 | NP_001304888.1 | 14 | 2116 | |||
| NM_152572.2 | NP_689785.1 | 12 | 1723 | |||
| XM_005272169.2 | XP_005272226.1 | 12 | 1262 | |||
| XM_006716965.2 | XP_006717028.1 | 10 | 1035 | |||
| XM_011518278.2 | XP_011516580.1 | 11 | 1103 | |||
| XM_017014308.1 | XP_016869797.1 | 6 | 785 | |||
| GenBank mRNA | AK057266.1 | - | 12 | 1196 | ||
| AK093446.1 | - | 12 | 1238 | |||
| AK292186.1 | - | 12 | 1208 | |||
| BC034776.1 | - | 12 | 1220 | 5175757 | ||
| BC050576.1 | - | 12 | 1723 | 5744517 | ||

AK8
Genomic Map