siRNA Details
| Gene Symbol: SAXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287049.1 | NP_001273978.1 | 4 | 1080 | ||
| NM_001287050.1 | NP_001273979.1 | 3 | 1119 | |||
| NM_153707.3 | NP_714918.2 | 4 | 1322 | |||
| XM_011517744.1 | XP_011516046.1 | 5 | 1402 | |||
| XM_011517745.1 | XP_011516047.1 | 6 | 1337 | |||
| XM_011517746.2 | XP_011516048.1 | 6 | 1258 | |||
| XM_011517747.2 | XP_011516049.1 | 3 | 918 | |||
| XM_017014314.1 | XP_016869803.1 | 6 | 1648 | |||
| XM_017014315.1 | XP_016869804.1 | 4 | 1035 | |||
| GenBank mRNA | AK057474.1 | - | 3 | 1114 | ||
| AK292245.1 | - | 4 | 1273 | |||
| BC033489.1 | - | 4 | 1250 | 5170510 | ||

SAXO1
Genomic Map