siRNA Details
| Gene Symbol: SAXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057474.1 | - | 2 | 523 | ||
| AK292245.1 | - | 2 | 479 | |||
| BC033489.1 | - | 2 | 456 | 5170510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAXO1| Gene Symbol: SAXO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057474.1 | - | 2 | 523 | ||
| AK292245.1 | - | 2 | 479 | |||
| BC033489.1 | - | 2 | 456 | 5170510 | ||