siRNA Details
| Gene Symbol: ZNF645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152577.3 | NP_689790.1 | 1 | 1143 | ||
| GenBank mRNA | AK098601.1 | - | 1 | 1115 | ||
| BC074910.2 | - | 1 | 1085 | 30915257 | ||
| BC126190.1 | - | 1 | 1085 | 8991906 | ||
| BC126192.1 | - | 1 | 1085 | 8991908 | ||
| BX103567.1 | - | 1 | 313 | |||
| GQ355336.1 | - | 1 | 1143 | |||

ZNF645
Genomic Map