siRNA Details
| Gene Symbol: ARHGAP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282607.1 | NP_001269536.1 | 11 | 1527 | ||
| NM_144967.3 | NP_659404.2 | 11 | 1763 | |||
| XM_006724729.1 | XP_006724792.1 | 10 | 1234 | |||
| XM_011531280.1 | XP_011529582.1 | 10 | 1251 | |||
| GenBank mRNA | AK054620.1 | - | 8 | 1123 | ||
| AK128203.1 | - | 11 | 1763 | |||
| AK296748.1 | - | 11 | 1505 | |||
| BC063790.1 | - | 11 | 1362 | 6196091 | ||

ARHGAP36
Genomic Map