siRNA Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031745.4 | NP_001026915.1 | 3 | 331 | ||
| NM_001267053.3 | NP_001253982.1 | 3 | 331 | |||
| NM_144968.3 | NP_659405.1 | 3 | 331 | |||
| XM_005261988.3 | XP_005262045.1 | 3 | 539 | |||
| XM_005261990.3 | XP_005262047.1 | 3 | 405 | |||
| GenBank mRNA | AK057345.1 | - | 3 | 297 | ||
| AK290346.1 | - | 3 | 281 | |||
| AK292871.1 | - | 3 | 286 | |||
| AK304179.1 | - | 3 | 331 | |||
| BC036767.1 | - | 3 | 246 | 5744127 | ||
| CF272701.1 | - | 4 | 400 | |||

RIBC1
Genomic Map