siRNA Details
| Gene Symbol: NKAPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027131.1 | 1 | 377 | |||
| GenBank mRNA | AK093895.1 | - | 1 | 343 | ||
| BC022359.1 | - | 1 | 317 | |||
| BC113730.1 | - | 1 | 316 | |||
| BC113732.1 | - | 1 | 316 | |||
| BX537594.1 | - | 1 | 369 | |||
| DB004517.1 | - | 1 | 379 | |||

NKAPP1
Genomic Map