siRNA Details
| Gene Symbol: WDFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020945.1 | NP_065996.1 | 15 | 2862 | ||
| XM_005270004.3 | XP_005270061.1 | 15 | 2977 | |||
| XM_011539986.2 | XP_011538288.2 | 15 | 2980 | |||
| XM_011539987.2 | XP_011538289.2 | 15 | 2983 | |||
| XM_011539988.2 | XP_011538290.1 | 16 | 3061 | |||
| XM_011539990.2 | XP_011538292.1 | 15 | 3008 | |||
| XM_011539991.2 | XP_011538293.1 | 15 | 3012 | |||
| XM_011539992.2 | XP_011538294.1 | 15 | 3012 | |||
| XM_017016463.1 | XP_016871952.1 | 16 | 2904 | |||
| XM_017016465.1 | XP_016871954.1 | 15 | 2989 | |||
| GenBank mRNA | AK127650.1 | - | 15 | 2871 | ||

WDFY4
Genomic Map