siRNA Details
| Gene Symbol: SLC5A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178498.3 | NP_848593.2 | 5 | 973 | ||
| XM_006718155.3 | XP_006718218.1 | 5 | 876 | |||
| XM_006718156.3 | XP_006718219.1 | 5 | 1212 | |||
| XM_011519920.2 | XP_011518222.1 | 6 | 453 | |||
| XM_011519921.2 | XP_011518223.1 | 5 | 1213 | |||
| XM_017017244.1 | XP_016872733.1 | 6 | 899 | |||
| GenBank mRNA | AY299482.1 | - | 5 | 682 | ||
| BC041454.1 | - | 5 | 442 | |||
| BC049207.1 | - | 5 | 945 | 5188979 | ||
| CN370367.1 | - | 1 | 82 | |||

SLC5A12
Genomic Map