siRNA Details
| Gene Symbol: CLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253750.1 | NP_001240679.1 | 1 | 213 | ||
| NM_001267701.1 | NP_001254630.1 | 1 | 213 | |||
| NM_172004.3 | NP_742001.1 | 1 | 213 | |||
| XM_011520574.2 | XP_011518876.1 | 1 | 256 | |||
| GenBank mRNA | AF518873.1 | - | 1 | 178 | ||
| BC093857.1 | - | 1 | 213 | 7939702 | ||
| BC093859.1 | - | 1 | 213 | 7939704 | ||
| DA619574.1 | - | 1 | 37 | |||

CLECL1
Genomic Map