siRNA Details
| Gene Symbol: CLECL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253750.1 | NP_001240679.1 | 2 | 506 | ||
| NM_001267701.1 | NP_001254630.1 | 2 | 506 | |||
| NM_172004.3 | NP_742001.1 | 2 | 506 | |||
| XM_011520574.2 | XP_011518876.1 | 2 | 549 | |||
| XM_017018885.1 | XP_016874374.1 | 3 | 386 | |||
| GenBank mRNA | AF518873.1 | - | 2 | 471 | ||
| AW237307.1 | - | 1 | 175 | |||
| BC093857.1 | - | 2 | 506 | 7939702 | ||
| BC093859.1 | - | 2 | 506 | 7939704 | ||
| BX103210.1 | - | 2 | 279 | |||
| DA619574.1 | - | 2 | 331 | |||

CLECL1
Genomic Map