siRNA Details
| Gene Symbol: TMTC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181783.3 | NP_861448.2 | 5 | 801 | ||
| XM_011537980.2 | XP_011536282.1 | 4 | 576 | |||
| GenBank mRNA | AJ697717.1 | - | 5 | 671 | ||
| AK074973.1 | - | 4 | 428 | |||
| AK290436.1 | - | 5 | 801 | |||
| AK302755.1 | - | 5 | 801 | |||
| AK310375.1 | - | 5 | 801 | |||
| AK310393.1 | - | 5 | 801 | |||
| BC117175.1 | - | 4 | 622 | 40125726 | ||
| BC117177.1 | - | 4 | 623 | 40125728 | ||
| BX538169.1 | - | 5 | 774 | |||
| CR749244.1 | - | 5 | 753 | |||
| CR749309.1 | - | 5 | 787 | |||
| CR933632.1 | - | 5 | 752 | |||

TMTC3
Genomic Map