siRNA Details
| Gene Symbol: STOML3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144033.1 | NP_001137505.1 | 6, 7 | 728 | ||
| NM_145286.2 | NP_660329.1 | 5, 6 | 644 | |||
| XM_017020400.1 | XP_016875889.1 | 3, 4 | 352 | |||
| GenBank mRNA | AK304159.1 | - | 6, 7 | 728 | ||
| AK313632.1 | - | 5, 6 | 616 | |||
| BC025760.1 | - | 5, 6 | 631 | 5204252 | ||

STOML3
Genomic Map