siRNA Details
| Gene Symbol: SYNE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152592.3 | NP_689805.3 | 14 | 2522 | ||
| XM_005267377.3 | XP_005267434.1 | 15 | 2619 | |||
| XM_006720063.3 | XP_006720126.1 | 15 | 2719 | |||
| XM_011536515.2 | XP_011534817.1 | 15 | 2654 | |||
| XM_017021056.1 | XP_016876545.1 | 14 | 2506 | |||
| GenBank mRNA | AK098471.1 | - | 9 | 1285 | ||
| AK131436.1 | - | 15 | 2372 | |||
| BC146604.1 | - | 15 | 2519 | 40123293 | ||

SYNE3
Genomic Map