siRNA Details
| Gene Symbol: MDGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113498.2 | NP_001106970.3 | 9 | 2128 | ||
| NM_182830.4 | NP_878250.2 | 9 | 1834 | |||
| XM_011536519.2 | XP_011534821.1 | 9 | 1983 | |||
| XM_011536525.2 | XP_011534827.1 | 6 | 1265 | |||
| XM_017021060.1 | XP_016876549.1 | 5 | 1094 | |||
| GenBank mRNA | AK309211.1 | - | 9 | 2131 | ||
| AY328482.1 | - | 3 | 761 | |||
| AY358125.1 | - | 8 | 1708 | |||
| AY369208.1 | - | 9 | 1924 | |||
| BC142947.1 | - | 9 | 1851 | |||

MDGA2
Genomic Map