siRNA Details
| Gene Symbol: C16orf82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145545.1 | NP_001139017.1 | 1 | 104 | ||
| GenBank mRNA | AF536240.1 | - | 1 | 295 | ||
| AK311574.1 | - | 1 | 46 | |||
| BC031257.1 | - | 1 | 77 | |||
| BC136370.1 | - | 1 | 287 | 9020357 | ||
| BC136382.1 | - | 1 | 287 | 9020369 | ||

C16orf82
Genomic Map