siRNA Details
| Gene Symbol: FBXO39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153230.2 | NP_694962.1 | 2 | 425 | ||
| XM_011523697.1 | XP_011521999.1 | 2 | 501 | |||
| XM_017024283.1 | XP_016879772.1 | 2 | 474 | |||
| GenBank mRNA | BC034782.1 | - | 2 | 405 | 5170425 | |
| BM554273.1 | - | 2 | 425 | |||

FBXO39
Genomic Map