siRNA Details
| Gene Symbol: KRT28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181535.3 | NP_853513.2 | 6, 7 | 1262 | ||
| GenBank mRNA | AJ564207.1 | - | 6, 7 | 1235 | ||
| AK129827.1 | - | 6, 7 | 1262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT28| Gene Symbol: KRT28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181535.3 | NP_853513.2 | 6, 7 | 1262 | ||
| GenBank mRNA | AJ564207.1 | - | 6, 7 | 1235 | ||
| AK129827.1 | - | 6, 7 | 1262 | |||