siRNA Details
| Gene Symbol: ZNF610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161425.1 | NP_001154897.1 | 6 | 1882 | ||
| NM_001161426.1 | NP_001154898.1 | 6 | 1769 | |||
| NM_001161427.1 | NP_001154899.1 | 5 | 1753 | |||
| NM_173530.2 | NP_775801.2 | 6 | 1738 | |||
| GenBank mRNA | AK093359.1 | - | 6 | 1738 | ||
| AK290888.1 | - | 6 | 1819 | |||
| AL831871.1 | - | 1 | 626 | |||
| BC039903.2 | - | 5 | 1697 | 6049284 | ||
| BC105967.1 | - | 6 | 1656 | 6154656 | ||
| DB577508.1 | - | 1 | 97 | |||

ZNF610
Genomic Map