siRNA Details
| Gene Symbol: ZNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301062.1 | NP_001287991.1 | 4 | 857 | ||
| NM_153608.2 | NP_705836.1 | 6 | 1192 | |||
| XM_011526574.2 | XP_011524876.1 | 6 | 1106 | |||
| XM_017026414.1 | XP_016881903.1 | 6 | 1078 | |||
| XM_017026415.1 | XP_016881904.1 | 5 | 1044 | |||
| GenBank mRNA | AK075062.1 | - | 5 | 919 | ||
| AK291575.1 | - | 6 | 1262 | |||
| BC014935.2 | - | 6 | 1201 | |||
| BC034032.1 | - | 1 | 241 | |||
| BC125068.1 | - | 1 | 918 | 40117544 | ||
| BC125069.1 | - | 4 | 787 | 40117550 | ||

ZNF114
Genomic Map