siRNA Details
| Gene Symbol: GBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320257.1 | NP_001307186.1 | 3 | 499 | ||
| NM_198460.2 | NP_940862.2 | 5 | 840 | |||
| XM_011540835.2 | XP_011539137.1 | 5 | 669 | |||
| GenBank mRNA | AK131356.1 | - | 5 | 824 | ||
| AK299408.1 | - | 4 | 475 | |||
| AK299466.1 | - | 3 | 316 | |||
| AK303987.1 | - | 5 | 584 | |||
| AK315834.1 | - | 5 | 658 | |||
| BC131713.1 | - | 5 | 618 | 40122722 | ||
| BX537949.1 | - | 5 | 659 | |||
| BX647907.1 | - | 5 | 657 | |||

GBP6
Genomic Map