siRNA Details
| Gene Symbol: FNDC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144937.1 | NP_001138409.1 | 7, 8 | 1374 | ||
| GenBank mRNA | AK093157.1 | - | 3, 4 | 700 | ||
| BC057762.1 | - | 3, 4 | 747 | |||
| BC130421.1 | - | 3, 4 | 700 | 40146452 | ||

FNDC7
Genomic Map