siRNA Details
| Gene Symbol: CCDC185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152610.2 | NP_689823.2 | 1 | 1377 | ||
| GenBank mRNA | AK093047.1 | - | 1 | 1333 | ||
| BC030026.1 | - | 1 | 1326 | 4822648 | ||
| BC031066.1 | - | 1 | 1324 | 5272765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC185| Gene Symbol: CCDC185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152610.2 | NP_689823.2 | 1 | 1377 | ||
| GenBank mRNA | AK093047.1 | - | 1 | 1333 | ||
| BC030026.1 | - | 1 | 1326 | 4822648 | ||
| BC031066.1 | - | 1 | 1324 | 5272765 | ||