siRNA Details
| Gene Symbol: WBP2NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152613.2 | NP_689826.2 | 3 | 325 | ||
| GenBank mRNA | AK128138.1 | - | 3 | 325 | ||
| AK129656.1 | - | 3 | 278 | |||
| BC022546.1 | - | 3 | 262 | 4811804 | ||
| BC022549.1 | - | 3 | 262 | 4815849 | ||
| CR456400.1 | - | 3 | 238 | |||
| DB079211.1 | - | 3 | 325 | |||

WBP2NL
Genomic Map