siRNA Details
| Gene Symbol: DAW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178821.1 | NP_849143.1 | 9 | 891 | ||
| XM_011510755.1 | XP_011509057.1 | 10 | 1500 | |||
| XM_017003490.1 | XP_016858979.1 | 8 | 906 | |||
| GenBank mRNA | AK097409.1 | - | 9 | 891 | ||
| AK098821.1 | - | 9 | 839 | |||
| AK127882.1 | - | 9 | 1448 | |||
| AK301861.1 | - | 10 | 891 | |||
| BC036377.2 | - | 9 | 871 | 4824989 | ||

DAW1
Genomic Map