siRNA Details
| Gene Symbol: ADGRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145168.1 | NP_001138640.1 | 7 | 958 | ||
| NM_001145169.1 | NP_001138641.1 | 6 | 804 | |||
| NM_001321971.1 | NP_001308900.1 | 6 | 1337 | |||
| NM_153835.3 | NP_722577.2 | 4 | 944 | |||
| NR_135916.1 | 4 | 944 | ||||
| XM_011532621.2 | XP_011530923.1 | 6 | 1289 | |||
| XM_011532622.2 | XP_011530924.1 | 6 | 1241 | |||
| XM_011532623.2 | XP_011530925.1 | 6 | 1233 | |||
| XM_017003505.1 | XP_016858994.1 | 6 | 1275 | |||
| XM_017003506.1 | XP_016858995.1 | 6 | 1531 | |||
| GenBank mRNA | AK293887.1 | - | 6 | 804 | ||
| AY140955.1 | - | 7 | 958 | |||
| AY358172.1 | - | 4 | 944 | |||

ADGRF3
Genomic Map