siRNA Details
| Gene Symbol: DNAJB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153330.4 | NP_699161.1 | 3 | 1720 | ||
| XM_006713519.3 | XP_006713582.1 | 2 | 2617 | |||
| GenBank mRNA | AK125946.1 | - | 2 | 2617 | ||
| AK126068.1 | - | 1 | 295 | |||
| BC029521.1 | - | 1 | 313 | 5296554 | ||
| BC050288.1 | - | 3 | 1720 | 5270636 | ||

DNAJB8
Genomic Map