siRNA Details
| Gene Symbol: SPATA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317799.1 | NP_001304728.1 | 9 | 1651 | ||
| NM_145207.2 | NP_660208.2 | 9 | 1654 | |||
| XM_005262783.4 | XP_005262840.1 | 9 | 1651 | |||
| XM_011531678.2 | XP_011529980.1 | 9 | 1651 | |||
| XM_011531679.2 | XP_011529981.1 | 9 | 1609 | |||
| XM_017007825.1 | XP_016863314.1 | 9 | 1650 | |||
| XM_017007826.1 | XP_016863315.1 | 9 | 1650 | |||
| XM_017007827.1 | XP_016863316.1 | 9 | 1650 | |||
| XM_017007828.1 | XP_016863317.1 | 9 | 1616 | |||
| XM_017007829.1 | XP_016863318.1 | 7 | 2274 | |||
| XM_017007830.1 | XP_016863319.1 | 9 | 1650 | |||
| GenBank mRNA | AF361489.1 | - | 9 | 1650 | ||
| AF479656.1 | - | 9 | 1624 | |||
| AK091384.1 | - | 9 | 1609 | |||
| AL832175.1 | - | 9 | 1647 | |||
| BC048217.1 | - | 9 | 1594 | 6052305 | ||

SPATA5
Genomic Map